Learn More
Chemglass Life Sciences Chemglass 250 mL Squibb separatory funnel, PYREX stopper #22, PTFE stopcock (CG-8622-250)
SDP
Catalog No. NC2099119
Encompass_Preferred
The Chemglass 250 mL Squibb (pear-shaped) separatory funnel is made from borosilicate (PYREX) glass and equipped with a PTFE stopcock and a #22 stopper for secure, chemical-resistant control of liquid-liquid phase separations. It is suited to routine extractions, solvent partitioning, and laboratory separations where durable glassware and inert valve materials are required.
  • Capacity: 250 mL
  • Stopper size: #22
  • Stopcock material: PTFE for chemical resistance
  • Body material: borosilicate (PYREX) glass
  • Shape: Squibb (pear-shaped)
  • Typical use: liquid-liquid extraction and phase separation
This item is not returnable. View return policy
Catalog No. NC2099119 Supplier Chemglass Life Sciences Supplier No. CG8622250
The Chemglass 250 mL Squibb (pear-shaped) separatory funnel is made from borosilicate (PYREX) glass and equipped with a PTFE stopcock and a #22 stopper for secure, chemical-resistant control of liquid-liquid phase separations. It is suited to routine extractions, solvent partitioning, and laboratory separations where durable glassware and inert valve materials are required.
  • Capacity: 250 mL
  • Stopper size: #22
  • Stopcock material: PTFE for chemical resistance
  • Body material: borosilicate (PYREX) glass
  • Shape: Squibb (pear-shaped)
  • Typical use: liquid-liquid extraction and phase separation