Learn More
Chemglass Life Sciences 250mL Single Neck RBF, 28/15 Socket Joint
SDP
Catalog No. 5000980654
Encompass_Preferred
Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.
Catalog No. 50-009-80654 Supplier Chemglass Life Sciences Supplier No. CG150805
Round bottom, HEAVY WALL single short neck flask with a spherical socket joint. Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.