Learn More
Polysciences, Inc. Soft Nail, 250 mL
SDP
Catalog No. NC9670470
Encompass_Preferred
Made for very hard complex keratinized hair, nail and skin samples common in Mohs Histology laboratories. Hair, nail and skin samples that have been processed tend to flex and turn becoming difficult to cut and hold on slides for staining.
Catalog No. NC9670470 Supplier Polysciences, Inc. Supplier No. 24775250
Made for very hard complex keratinized hair, nail and skin samples common in Mohs Histology laboratories. Hair, nail and skin samples that have been processed tend to flex and turn becoming difficult to cut and hold on slides for staining.