Learn More
Bellco Glass, Inc. 250mL Square shape PETG Media Bottles, Sterile, 6/pk, 48/cs
SDP
Catalog No. NC3865166
Encompass_Preferred
250mL PETG Square Media Bottles, Sterile, 6/pk, 48/cs. It is mainly used for storing and sampling active pharmaceutical ingredients, bulk intermediates, and also for preparing, storing buffers, culture solutions or long-term storage of pH sensitive liquids. Tips: Can't autoclave. It can be used to storage reagents but cannot be used as heating vessels. PETG Bottles are Suitable for handling and reagents and medium storing critical(protein/ vaccine etc.PET Bottles are used for storage of culture medium, serum, buffer and long-term storage of PH-sensitive liquid. Excellent physical and mechanical properties over a wide temperature range.Temperature Range: -80 oC to 50 oC.
Catalog No. NC3865166 Supplier Bellco Glass, Inc. Supplier No. 451400432/SQ1026866
250mL PETG Square Media Bottles, Sterile, 6/pk, 48/cs. It is mainly used for storing and sampling active pharmaceutical ingredients, bulk intermediates, and also for preparing, storing buffers, culture solutions or long-term storage of pH sensitive liquids. Tips: Can't autoclave. It can be used to storage reagents but cannot be used as heating vessels. PETG Bottles are Suitable for handling and reagents and medium storing critical(protein/ vaccine etc.PET Bottles are used for storage of culture medium, serum, buffer and long-term storage of PH-sensitive liquid. Excellent physical and mechanical properties over a wide temperature range.Temperature Range: -80 oC to 50 oC.