Learn More
Chemglass Life Sciences Bottle, 250mL, Storage, 24/40 Outer Joint, Single Neck
SDP
Catalog No. CG112601
Encompass_Preferred
Bottle, 250mL, Storage, 24/40 Outer Joint, Single Neck. Bottle has a standard taper outer joint. Supplied without stopper. Bottles are available amberized for protection of light sensitive materials.
Catalog No. CG112601 Supplier Chemglass Life Sciences Supplier No. CG112601
Bottle, 250mL, Storage, 24/40 Outer Joint, Single Neck. Bottle has a standard taper outer joint. Supplied without stopper. Bottles are available amberized for protection of light sensitive materials.