Learn More
Chemglass Life Sciences 250mL Flask, Storage, 24/40 Outer Joint, Airfree, Schlenk
SDP
Catalog No. AF052207
Encompass_Preferred
250mL Flask, Storage, 24/40 Outer Joint, Airfree, Schlenk. Round bottom storage flask with a standard taper outer joint on the sidearm. The 50, 100 and 250mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 500 and 1000mL sizes have a 0-8mm valve size.
Catalog No. AF052207 Supplier Chemglass Life Sciences Supplier No. AF052207
250mL Flask, Storage, 24/40 Outer Joint, Airfree, Schlenk. Round bottom storage flask with a standard taper outer joint on the sidearm. The 50, 100 and 250mL sizes have a 0-4mm Chem-Vac™ Chem-Cap valve while the 500 and 1000mL sizes have a 0-8mm valve size.