Learn More
Ace Glass, Inc. Trap, rotary evaporator, self-washing, 250mL, 24/40 top & bottom
SDP
Catalog No. NC0814276
Encompass_Preferred
250mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 joints, top and bottom.
Catalog No. NC0814276 Supplier Ace Glass, Inc. Supplier No. 670320
250mL self-washing, rotary evaporator trap designed by Dr. William Boulanger. Design prevents blow-through and enables continuous addition with rotary evaporators. Trap has 24/40 joints, top and bottom.