Learn More
Chemglass Life Sciences 250mL Volumetric Flask, HW, Bubble Neck, Large Numbers, Flat Bottom, Class A, TC, #19 Stopper
SDP
Catalog No. 501531198
Encompass_Preferred
250mL Volumetric Flask, Heavy Wall, Bubble Neck, Large Numbers, Flat Bottom, Class A, TC, #19 Stopper.
Catalog No. 50-153-1198 Supplier Chemglass Life Sciences Supplier No. CG1626250
250mL Volumetric Flask, Heavy Wall, Bubble Neck, Large Numbers, Flat Bottom, Class A, TC, #19 Stopper.