Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chemglass Life Sciences Chemglass 250 mL Beaker, Low Form (Griffin), Pyrex, Double Scale
SDP

Catalog No. NC2217093
Encompass_Preferred

250 mL low-form Griffin beaker made from Pyrex borosilicate glass for general laboratory mixing, heating, and reagent handling. The beaker provides approximate volume markings and a large marking spot for easy measurement and labeling.

  • Capacity: 250 mL
  • Material: Borosilicate glass
  • Form: Low form (Griffin)
  • Graduations: Double graduated scale for approximate volume
  • Marking spot: Extra-large marking spot for labeling
  • Dimensions (approx.): 68 x 90 mm (OD x height)
  • Standards: Meets ASTM E960, Type I
  • Construction: Uniform wall thickness for thermal shock resistance and mechanical strength

This item is not returnable. View return policy
Catalog No. NC2217093 Supplier Chemglass Life Sciences Supplier No. CG8048250
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

250 mL low-form Griffin beaker made from Pyrex borosilicate glass for general laboratory mixing, heating, and reagent handling. The beaker provides approximate volume markings and a large marking spot for easy measurement and labeling.

  • Capacity: 250 mL
  • Material: Borosilicate glass
  • Form: Low form (Griffin)
  • Graduations: Double graduated scale for approximate volume
  • Marking spot: Extra-large marking spot for labeling
  • Dimensions (approx.): 68 x 90 mm (OD x height)
  • Standards: Meets ASTM E960, Type I
  • Construction: Uniform wall thickness for thermal shock resistance and mechanical strength

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.