Learn More
Bellco Glass, Inc. 250mL Wide Mouth Bottle
SDP
Catalog No. 502330370
Encompass_Preferred
CELLTREAT Polypropylene Wide Mouth BottleSpecifications
Round shape and easypour wide mouth design Excellent chemical resistance and durable rigid walls Ideal for specimen and chemical storage Linerless leak proof screw top closure Assembled bottle and cap Size 250mL Pack Description 6/Bag Case Quantity 24
Catalog No. 50-233-0370 Supplier Bellco Glass, Inc. Supplier No. 229796000
CELLTREAT Polypropylene Wide Mouth BottleSpecifications
Round shape and easypour wide mouth design Excellent chemical resistance and durable rigid walls Ideal for specimen and chemical storage Linerless leak proof screw top closure Assembled bottle and cap Size 250mL Pack Description 6/Bag Case Quantity 24