Learn More
Chemglass Life Sciences 250mL 3-Neck Round Bottom Flask (RBF) , 1-CN 29/26 Outer, 2-SN 29/26 Outer
SDP
Catalog No. CG152290
Encompass_Preferred
250mL 3-Neck Round Bottomed Flask, 1-CN 29/26 Outer, 2-SN 29/26 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in clamping of the flask.
Catalog No. CG152290 Supplier Chemglass Life Sciences Supplier No. CG152290
250mL 3-Neck Round Bottomed Flask, 1-CN 29/26 Outer, 2-SN 29/26 Outer. Round bottom, HEAVY WALL three neck flask with standard taper outer joints on the center neck and vertical side necks. Flasks with 29/26 center necks have a groove tooled just below the grind to aid in clamping of the flask.