Learn More
Ace Glass, Inc. 250MM COLUMN SILVERED 250MM COLUMN SILVERED
SDP
Catalog No. 50975421
Encompass_Preferred
250mm silvered Hempel distillation column is vacuum jacketed and has 24/40 standard taper joints. Effective length is 250mm.
Catalog No. 50-975-421 Supplier Ace Glass, Inc. Supplier No. 779312
250mm silvered Hempel distillation column is vacuum jacketed and has 24/40 standard taper joints. Effective length is 250mm.