Learn More
Ace Glass, Inc. 250mm column unsilvered see p95, 6620
SDP
Supplier:  Ace Glass, Inc. 779304
Encompass_Preferred
250mm unsilvered Hempel distillation column is vacuum jacketed and has 24/40 standard taper joints. Effective length is 250mm.
Catalog No. 50-975-420