Learn More
Chemglass Life Sciences Chemglass 250 μL Automation Clear Pipette Tips for Beckman Systems, Sterile - 96 tips/rack, 10 racks/pack (50 racks/case)
SDP
Catalog No. 502733231
Encompass_Preferred
250 μL automation clear pipette tips designed for high-throughput liquid handling and molecular biology workflows. Manufactured from medical-grade polypropylene and supplied sterile to ensure sample integrity.
  • 250 μL nominal volume.
  • Sterile, electron beam sterilized.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Medical-grade polypropylene construction.
  • DNase/RNase free.
  • Suitable for high-throughput genomics, proteomics, and immunoassay workflows.
Catalog No. 50-273-3231 Supplier Chemglass Life Sciences Supplier No. CLS3499A501
Add to Cart
Add to Cart
250 μL automation clear pipette tips designed for high-throughput liquid handling and molecular biology workflows. Manufactured from medical-grade polypropylene and supplied sterile to ensure sample integrity.
  • 250 μL nominal volume.
  • Sterile, electron beam sterilized.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Medical-grade polypropylene construction.
  • DNase/RNase free.
  • Suitable for high-throughput genomics, proteomics, and immunoassay workflows.