Learn More
Chemglass Life Sciences Chemglass 250 μL Automation Clear Pipette Tips for Agilent Systems, Sterile, 96/rack
SDP
Catalog No. 502733223
Encompass_Preferred
Chemglass 250 μL automation clear tips for Agilent systems are sterile, low-retention polypropylene pipette tips designed for high-throughput automated liquid handling. Pre-racked packaging supports efficient robot loading and reduces contamination risk.
  • Volume: 250 μL
  • Compatible system: Agilent automated liquid handlers
  • Packaging: 96 units per rack, 10 racks per pack, 50 racks per case
  • Material: medical-grade polypropylene
  • Sterility: electron beam sterilized
  • Free of DNase and RNase
  • Manufactured in a 100,008-grade clean room
  • Low-retention surface to reduce liquid hanging on the wall
Catalog No. 50-273-3223 Supplier Chemglass Life Sciences Supplier No. CLS3499A703
Add to Cart
Add to Cart
Chemglass 250 μL automation clear tips for Agilent systems are sterile, low-retention polypropylene pipette tips designed for high-throughput automated liquid handling. Pre-racked packaging supports efficient robot loading and reduces contamination risk.
  • Volume: 250 μL
  • Compatible system: Agilent automated liquid handlers
  • Packaging: 96 units per rack, 10 racks per pack, 50 racks per case
  • Material: medical-grade polypropylene
  • Sterility: electron beam sterilized
  • Free of DNase and RNase
  • Manufactured in a 100,008-grade clean room
  • Low-retention surface to reduce liquid hanging on the wall