Learn More
Chemglass Life Sciences 250uL Automation Low-retention Clear Tips for Agilent System, Sterile, 96/rack, 10 racks/pk, 50 racks/cs
SDP
Catalog No. 502733227
Encompass_Preferred
NEST automation tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals. Compatible with Agilent Systems.Low-retention tips are and excellent choice for transferring viscous liquid, which can effectively reduce the phenomenon of liquid hanging on the wall and make the experiment more accurate.• Medical-grade PP • Dnase/Rnase free • Electron beam sterilization • Produced in 100,008 grade clean room
Catalog No. 50-273-3227 Supplier Chemglass Life Sciences Supplier No. CLS3499A603
Add to Cart
Add to Cart
NEST automation tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals. Compatible with Agilent Systems.Low-retention tips are and excellent choice for transferring viscous liquid, which can effectively reduce the phenomenon of liquid hanging on the wall and make the experiment more accurate.• Medical-grade PP • Dnase/Rnase free • Electron beam sterilization • Produced in 100,008 grade clean room