Learn More
Chemglass Life Sciences Chemglass Automation Low-Retention 250 μL Filter Pipette Tips for Agilent Systems - Sterile, Aerosol-Resistant, Rack-Nested (96 tips/rack)
SDP
Catalog No. 502733226
Encompass_Preferred
Sterile low-retention 250 μL filter pipette tips designed for automated liquid-handling systems. These aerosol-resistant tips reduce sample carryover and minimize sample adhesion for improved accuracy in high-throughput workflows.
  • 250 μL nominal volume.
  • Low-retention design reduces sample adhesion.
  • Aerosol-resistant filter minimizes carryover.
  • Sterile; electron beam sterilized.
  • RNase and DNase free.
  • Medical-grade polypropylene construction; clear.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Suitable for high-throughput automated pipetting and viscous liquids.
Catalog No. 50-273-3226 Supplier Chemglass Life Sciences Supplier No. CLS3499A604
Add to Cart
Add to Cart
Sterile low-retention 250 μL filter pipette tips designed for automated liquid-handling systems. These aerosol-resistant tips reduce sample carryover and minimize sample adhesion for improved accuracy in high-throughput workflows.
  • 250 μL nominal volume.
  • Low-retention design reduces sample adhesion.
  • Aerosol-resistant filter minimizes carryover.
  • Sterile; electron beam sterilized.
  • RNase and DNase free.
  • Medical-grade polypropylene construction; clear.
  • 96 tips per rack; 10 racks per pack; 50 racks per case.
  • Suitable for high-throughput automated pipetting and viscous liquids.