Learn More
Alkali Scientific 250uL Beckman Biomek FX/NX Mul
SDP
Catalog No. 503029129
Encompass_Preferred
Alkali Scientificdesigns its P250 black conductive automation tips to provide seamless compatibility with Beckman Coulter Biomek i-Series (i5 and i7) as well as previous NX/FX/FXP robotic liquid handlers No separate labware definition is required and existing automation control software protocols do not need modification The Alkali ScientificP250 (250 L) tip is one of the most popular choices in the 96-format category Known for its excellent performance at sub-microliter volumes it offers outstanding precision and accuracy Its volume capacity and length provide flexibility in protocol design These tips are compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek systems Manufactured from the highest quality virgin polypropylene Alkali Scientificensures low liquid retention in its P250 black conductive tips through advanced polishing techniques without the use of releasing agents or additives Specifications Working volume 250 00 L Well f
Catalog No. 50-302-9129 Supplier Alkali Scientific Supplier No. FXBS250CS
Alkali Scientificdesigns its P250 black conductive automation tips to provide seamless compatibility with Beckman Coulter Biomek i-Series (i5 and i7) as well as previous NX/FX/FXP robotic liquid handlers No separate labware definition is required and existing automation control software protocols do not need modification The Alkali ScientificP250 (250 L) tip is one of the most popular choices in the 96-format category Known for its excellent performance at sub-microliter volumes it offers outstanding precision and accuracy Its volume capacity and length provide flexibility in protocol design These tips are compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek systems Manufactured from the highest quality virgin polypropylene Alkali Scientificensures low liquid retention in its P250 black conductive tips through advanced polishing techniques without the use of releasing agents or additives Specifications Working volume 250 00 L Well f