Alkali Scientificdesigns its P250 automation tips to be fully compatible with Beckman Coulter Biomek i-Series (i5 and i7) as well as earlier NX/FX/FXP robotic liquid handlers These tips work seamlessly without the need for a separate labware definition or modifications to existing automation control software protocols The Alkali ScientificP250 (250 L) tip is one of the most popular options in the 96-format category known for its excellent precision and accuracy in sub-microliter performance Its volume capacity and length provide versatility for designing a variety of protocols It is compatible with all 96 multi-channel heads on Biomek i-Series (i5 and i7) NX/FX/FXP and Multimek systems Alkali Scientificmanufactures its P250 tips using the highest quality virgin polypropylene Through advanced polishing techniques we ensure minimal liquid retention without the use of releasing agents or additives Specifications Working volume 250 00 L Well format 96 96 384 plate accessible Head