Learn More
Labsource Inc 250 µL Beckman Non-Barrier Tips, Non-Sterile, 100% Premium Polypropylene
SDP
Catalog No. NC3095880
Encompass
These 250 µL Beckman non-barrier pipette tips provide precise liquid handling in automated lab systems. Their premium polypropylene makeup ensures high performance and purity, making them ideal for sensitive applications. With rigorous quality control and a leak-proof design, they stand out for consistent accuracy, making them a reliable choice for high-throughput liquid handling, genomics, and drug discovery.
Catalog No. NC3095880 Supplier Labsource Inc Supplier No. P21282
These 250 µL Beckman non-barrier pipette tips provide precise liquid handling in automated lab systems. Their premium polypropylene makeup ensures high performance and purity, making them ideal for sensitive applications. With rigorous quality control and a leak-proof design, they stand out for consistent accuracy, making them a reliable choice for high-throughput liquid handling, genomics, and drug discovery.