Learn More
Alkali Scientific 250uL CyBio-Style Analytik Jen
SDP
Catalog No. 503026423
Encompass_Preferred
Enhance your liquid handling processes with our 250 L Clear Robotic Automation Tips designed for flawless compatibility with CyBio FeliX Well Flex and Vario Liquid Workstations Each box contains 768 tips (3840 tips per case) providing a dependable supply for your high-throughput applications Key Features Precision Engineering Specifically crafted for the Cybi-Well Family including Cybi-Well Cybi-Well Vario and Cybi-WellFlex these tips ensure consistent accurate liquid handling for all your automated workflows Low Retention Design Our low retention technology reduces sample loss ensuring optimal performance and reproducibility in every experiment Sterile and Clean Manufactured in a 100000-class cleanroom (Class 8) these tips are DNase RNase and pyrogen free Each batch is sterilized using E-beam technology and verified by SGS for your assurance Versatile Application Perfect for a range of liquid handling tasks these tips are designed for compatibility with a 96-well head/tip format
Catalog No. 50-302-6423 Supplier Alkali Scientific Supplier No. CBFT10250
Enhance your liquid handling processes with our 250 L Clear Robotic Automation Tips designed for flawless compatibility with CyBio FeliX Well Flex and Vario Liquid Workstations Each box contains 768 tips (3840 tips per case) providing a dependable supply for your high-throughput applications Key Features Precision Engineering Specifically crafted for the Cybi-Well Family including Cybi-Well Cybi-Well Vario and Cybi-WellFlex these tips ensure consistent accurate liquid handling for all your automated workflows Low Retention Design Our low retention technology reduces sample loss ensuring optimal performance and reproducibility in every experiment Sterile and Clean Manufactured in a 100000-class cleanroom (Class 8) these tips are DNase RNase and pyrogen free Each batch is sterilized using E-beam technology and verified by SGS for your assurance Versatile Application Perfect for a range of liquid handling tasks these tips are designed for compatibility with a 96-well head/tip format