Learn More
Trajan Scientific L-MARK Brand 250l Syringe, FN, GT, 22g, NDL Length 51mm, Pt. Style 3, Plunger Tip PE, 6 Pack
Catalog No. 50635353
Encompass_Preferred
L-MARK Brand Syringe, 250uL, FN, GT, PE, 22g, Pt. 3 (Barrel 6.6mm), 6 Pack; Replacement Plunger: LMK.2636808; Syringe Adapter: PAL.SA250A Made in Germany.
Catalog No. 50-635-353 Supplier Trajan Scientific Supplier No. LMK.26208206P
L-MARK Brand Syringe, 250uL, FN, GT, PE, 22g, Pt. 3 (Barrel 6.6mm), 6 Pack; Replacement Plunger: LMK.2636808; Syringe Adapter: PAL.SA250A Made in Germany.