Learn More
Trajan Scientific L-MARK Brand 250l Syringe, FN, GT, 22g, NDL Length 51mm, Pt. Style 3, Plunger Tip PTFE, Single
Catalog No. 50635355
Encompass_Preferred
L-MARK Brand Syringe, 250uL, FN, GT, PTFE, 22g, Pt. 3, (Barrel 6.6mm); Syringe Adapter: PAL.SA250A Made in Germany.
Catalog No. 50-635-355 Supplier Trajan Scientific Supplier No. LMK.2620821
L-MARK Brand Syringe, 250uL, FN, GT, PTFE, 22g, Pt. 3, (Barrel 6.6mm); Syringe Adapter: PAL.SA250A Made in Germany.