Learn More
Bellco Glass, Inc. 250µL Pipette Tips, MLA, Stacked Dense Rack, Non-sterile
SDP
Catalog No. 502330374
Encompass_Preferred
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time. Stacked Rack•Non-sterile only•Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved•200 tips/layer •For Small volume (250 mL) tips - 5 layers - 1000 tips •For Large volume (1250 mL) tips - 3 layers - 600 tips
Catalog No. 50-233-0374 Supplier Bellco Glass, Inc. Supplier No. 451900175
A superior fit for your MLA & Ovation Pipettes. Use genuine VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time. Stacked Rack•Non-sterile only•Tips and racks are manufactured from 100% virgin polypropylene and can be autoclaved•200 tips/layer •For Small volume (250 mL) tips - 5 layers - 1000 tips •For Large volume (1250 mL) tips - 3 layers - 600 tips