Learn More
MSC 3M 7000088503 3" Wide x 180' Long x 6.5 mil Thick Brown Masking Tape Paper Backing, Rubber Adhesive, 35 Lb/In Tensile Strength, 300°F Max, Series 2517
Catalog No. 501297602
Encompass
3M 7000088503 3" Wide x 180' Long x 6.5 mil Thick Brown Masking Tape Paper Backing, Rubber Adhesive, 35 Lb/In Tensile Strength, 300°F Max, Series 2517
Catalog No. 50-129-7602 Supplier MSC Supplier No. 04582938
3M 7000088503 3" Wide x 180' Long x 6.5 mil Thick Brown Masking Tape Paper Backing, Rubber Adhesive, 35 Lb/In Tensile Strength, 300°F Max, Series 2517