Learn More
MSC Three M Flatback Tape 2525, 1 in, 1 Roll
Catalog No. 501149442
Encompass
3M Performance Flatback Tape 2525 is a dependable tape suitable for splicing and paint masking applications that require adhesion in high-temperature bake cycles. It features a robust, smooth paper backing that resists high temperatures and moisture, and can be torn by hand.
  • Designed for high-temperature splicing and paint masking
  • Withstands bake cycles up to 300°F (149°C) for up to one hour
  • Natural rubber adhesive provides dependable tack and holding power
  • Offers quick and simple roll starting and roll closure
  • Durable backing can be torn by hand, yet suitable for machine dispensing
  • Resistant to moist conditions for lasting hold
  • Orange color provides high visibility and easy identification
Catalog No. 50-114-9442 Supplier MSC Supplier No. 04583035
3M Performance Flatback Tape 2525 is a dependable tape suitable for splicing and paint masking applications that require adhesion in high-temperature bake cycles. It features a robust, smooth paper backing that resists high temperatures and moisture, and can be torn by hand.
  • Designed for high-temperature splicing and paint masking
  • Withstands bake cycles up to 300°F (149°C) for up to one hour
  • Natural rubber adhesive provides dependable tack and holding power
  • Offers quick and simple roll starting and roll closure
  • Durable backing can be torn by hand, yet suitable for machine dispensing
  • Resistant to moist conditions for lasting hold
  • Orange color provides high visibility and easy identification