Learn More
Key Scientific Products 253OCARE 50/PK
SDP
Catalog No. NC9987343
Encompass_Preferred
PROTECT VIAL,1.5 mL freezer vial,mixed each color,50 per box,consists of 20-25 porous ceramic beads suspended in a unique cryopeservative fluid,store reserve organisms like bacteria,fungi,yeasts,anaerobes, Neisseria,Haemophilus for years
Catalog No. NC9987343 Supplier Key Scientific Products Supplier No. KS70MI50
PROTECT VIAL,1.5 mL freezer vial,mixed each color,50 per box,consists of 20-25 porous ceramic beads suspended in a unique cryopeservative fluid,store reserve organisms like bacteria,fungi,yeasts,anaerobes, Neisseria,Haemophilus for years