Learn More
Waters Corp Waters Corporation Performance Maintenance Kit, 2545 Quaternary Gradient Module, HPLC, Annual
Catalog No. 50791588 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters™ Performance Maintenance Kit for the 2545 Quaternary Gradient Module contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains all necessary parts for optimal performance
  • Includes solvent filters
  • Includes diffuser filters
  • Includes check valve cartridges
  • Includes plunger seals
  • Includes o-rings
  • Includes sapphire plungers
  • Includes seals for plunger wash
  • Includes outlet filter
  • Includes vent valve rotor seal
  • Designed for annual replacement
Catalog No. 50-791-588 Supplier Waters Corp Supplier No. 201000199
The Waters™ Performance Maintenance Kit for the 2545 Quaternary Gradient Module contains all the Waters Quality Parts™ necessary to keep your instrument operating at peak performance on an annual basis.
  • Contains all necessary parts for optimal performance
  • Includes solvent filters
  • Includes diffuser filters
  • Includes check valve cartridges
  • Includes plunger seals
  • Includes o-rings
  • Includes sapphire plungers
  • Includes seals for plunger wash
  • Includes outlet filter
  • Includes vent valve rotor seal
  • Designed for annual replacement