Learn More
Cayman Chemical 25B-NBF (hydrochloride), Purity: ≥98%, CAS: 1539266-17-5, 10 mg
SDP
Catalog No. 502972077
Encompass_Preferred
An N-fluorobenzyl derivative of the phenethylamine hallucinogen 2C-I that binds human 5-HT2A receptors and rat 5-HT2C receptors with pKi values of 8.57 and 7.73, respectively; intended for forensic and research applications
Catalog No. 50-297-2077 Supplier Cayman Chemical Supplier No. 1596710 MG
An N-fluorobenzyl derivative of the phenethylamine hallucinogen 2C-I that binds human 5-HT2A receptors and rat 5-HT2C receptors with pKi values of 8.57 and 7.73, respectively; intended for forensic and research applications