Learn More
Cayman Chemical 25C-NBOHhydrochlorIde 10mg
SDP
Catalog No. 502968567
Encompass_Preferred
A derivative of 2C-C having an N-(2-hydroxybenzyl) addition at the amine; a potent agonist of serotonin receptors that shows selectivity for 5-HT2A over 5-HT2C (EC50s = ~ 0.40 and 15 nM, respectively); intended for forensic and research applications
Catalog No. 50-296-8567 Supplier Cayman Chemical Supplier No. 1481510 MG
A derivative of 2C-C having an N-(2-hydroxybenzyl) addition at the amine; a potent agonist of serotonin receptors that shows selectivity for 5-HT2A over 5-HT2C (EC50s = ~ 0.40 and 15 nM, respectively); intended for forensic and research applications