Learn More
Alkali Scientific 25cm² Cell Culture Flasks, Vent Cap, Non-Treated, Sterile, 10/pk, 200/cs
SDP
Catalog No. 502336176
Encompass_Preferred
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing, clear graduations, and clear lot number for batch traceability. 200 per case.
Catalog No. 50-233-6176 Supplier Alkali Scientific Supplier No. TL31
These Cell Culture Flasks are non-treated, vented and sterile. They come packaged in zip bags and are high clarity, 100% virgin polystyrene. They are stackable, sterilized by E-Beam, non-pyrogenic, and DNase/RNase free. They have frosted writing, clear graduations, and clear lot number for batch traceability. 200 per case.