Learn More
Bellco Glass, Inc. 25cm2 Cell Culture Flask, Vent Cap, TC, sterile 10/pk, 200/cs
SDP
Catalog No. 502330819
Encompass_Preferred
25cm2 Cell Culture Flask, Vent Cap, TC, sterile, Maximum Working Volume 50mL, 10/pk, 200/cs. cell culture flasks are available in cell growth areas ranging from 25cm2 to 225cm2. Tissue culture treated or non-treated flasks available with a vent cap or plug seal cap to meet your needs. • Made of high clarity, 100% virgin polystyrene.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Frosted writing and clear graduations.• Notched bottom for slip free stacking.• Clear lot number for batch traceability.• Packaged in sterile, zip-sealable bags
Catalog No. 50-233-0819 Supplier Bellco Glass, Inc. Supplier No. 451400063
25cm2 Cell Culture Flask, Vent Cap, TC, sterile, Maximum Working Volume 50mL, 10/pk, 200/cs. cell culture flasks are available in cell growth areas ranging from 25cm2 to 225cm2. Tissue culture treated or non-treated flasks available with a vent cap or plug seal cap to meet your needs. • Made of high clarity, 100% virgin polystyrene.• Sterilized by E-beam, SAL=10-6.• Non-Pyrogenic, DNase/Rnase free.• Frosted writing and clear graduations.• Notched bottom for slip free stacking.• Clear lot number for batch traceability.• Packaged in sterile, zip-sealable bags