Learn More
Thomas Scientific 25cm2 Tissue Culture Flask - Vent Cap, Sterile
SDP
Catalog No. 501461474
Encompass_Preferred
CELLTREAT Tissue Culture flasks are manufactured under the most stringent conditions and include a 100% integrity test to assure the reliability with your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used for CELLTREAT products provides a negatively charged hydrophilic surface and results in a consistent cell attachment surface. Growth surfaces are flat and free from striation to maximize usable growth area and transparency. Flasks are available in four different sizes; 12.5cm 2 , 25cm 2 , 75cm 2 and 182cm 2 . Two different cap styles are available. Plug seal caps can provide a closed system or allow gas exchange with a one quarter turn of the cap. Vent caps contain a 0.22 µm filter to allow gas exchange and minimize the risk of cross contamination. Innovative angled neck
This item is not returnable. View return policy
Catalog No. 50-146-1474 Supplier Thomas Scientific Supplier No. 1228K52
CELLTREAT Tissue Culture flasks are manufactured under the most stringent conditions and include a 100% integrity test to assure the reliability with your tissue and cell culture work. Class 100,000 clean room, USP class VI virgin polystyrene, and ISO 9001:2000 quality management systems all contribute to the consistent tissue and cell culture performance of our products. The tissue culture surface treatment used for CELLTREAT products provides a negatively charged hydrophilic surface and results in a consistent cell attachment surface. Growth surfaces are flat and free from striation to maximize usable growth area and transparency. Flasks are available in four different sizes; 12.5cm 2 , 25cm 2 , 75cm 2 and 182cm 2 . Two different cap styles are available. Plug seal caps can provide a closed system or allow gas exchange with a one quarter turn of the cap. Vent caps contain a 0.22 µm filter to allow gas exchange and minimize the risk of cross contamination. Innovative angled neck