Learn More
Perkin Elmer US LLC 25-Element Solution, 5% HNO3, 100 µg/mL, 100 mL
Catalog No. 502117323
Encompass_Preferred
Details Concentration Element 100 µg/mL Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Se, Sr, Tl, U, V, Zn
Catalog No. 50-211-7323 Supplier Perkin Elmer US LLC Supplier No. N9308543
Details Concentration Element 100 µg/mL Ag, As, B, Ba, Be, Bi, Ca, Cd, Co, Fe, K, Li, Mg, Mn, Na, Ni, P, Pb, Se, Sr, Tl, U, V, Zn