Learn More
Trajan Scientific 25F-CTC-GT-5/0.47C
SDP
Catalog No. 502530347
Encompass_Preferred
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 25 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Needle Point Style: Cone, Description: Gas tight, Length: 2 in., 50 mm
Catalog No. 50-253-0347 Supplier Trajan Scientific Supplier No. 003700
SGE PAL Autosampler Syringes with 50 mm Fixed Cone Tip Needle, Volume: 25 uL, Connection Type: Fixed, For Use With: CTC, LEAP, CombiPAL, GC PAL, Gas chromatography, Length Needle: 50 mm, 1.96 in., Needle Gauge: 26 G, Needle Point Style: Cone, Description: Gas tight, Length: 2 in., 50 mm