Learn More
Trajan Scientific 25F-RTC/RSH-GT-5.7/0.63C 25UL
SDP
Catalog No. 502530345
Encompass_Preferred
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 25F-RTC/RSH-GT-5.7/0.63C, Volume: 25 uL, Connection Type: Fixed
Catalog No. 50-253-0345 Supplier Trajan Scientific Supplier No. 003862
Trajan Autosampler Syringes With Fixed Needle for Thermo Scientific GCs, Diameter Outer Needle: 0.02 in., 0.63 mm, Needle Gauge: 23 G, Length Needle: 2.2 in., 57 mm, Model: 25F-RTC/RSH-GT-5.7/0.63C, Volume: 25 uL, Connection Type: Fixed