Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester, 98%, [847818-74-0], M2011 208.07 C10H17BN2O2

Catalog No. 5038487
Encompass_Preferred

Compound: 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

CAS#: 847818-74-0

MDL#: not listed

Background: An N-methylated pyrazole boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It provides direct access to substituted pyrazole-containing scaffolds in drug discovery synthesis.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-384-87 Supplier Frontier Specialty Chemicals Supplier No. M201125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Methyl-1H-pyrazole-5-boronic acid pinacol ester

CAS#: 847818-74-0

MDL#: not listed

Background: An N-methylated pyrazole boronic ester used as a Suzuki-Miyaura coupling partner in pharmaceutical research. It provides direct access to substituted pyrazole-containing scaffolds in drug discovery synthesis.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.