Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 1-Boc-pyrrole-2-boronic acid, 98%, [135884-31-0], B5969 211.02 C9H14BNO4

Catalog No. 50981679
Encompass_Preferred

Compound: 1-Boc-pyrrole-2-boronic acid

CAS#: 135884-31-0

MDL#: MFCD01318939

Background: 1-Boc-pyrrole-2-boronic acid is a protected heteroaryl boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions. The Boc protecting group allows selective deprotection and further functionalization of the pyrrole ring, making it useful in pharmaceutical synthesis. It has also been applied in the development of sensors for detecting sugars and other biomolecules.

Catalog No. 50-981-679 Supplier Frontier Specialty Chemicals Supplier No. B596925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Boc-pyrrole-2-boronic acid

CAS#: 135884-31-0

MDL#: MFCD01318939

Background: 1-Boc-pyrrole-2-boronic acid is a protected heteroaryl boronic acid used as a coupling partner in Suzuki-Miyaura cross-coupling reactions. The Boc protecting group allows selective deprotection and further functionalization of the pyrrole ring, making it useful in pharmaceutical synthesis. It has also been applied in the development of sensors for detecting sugars and other biomolecules.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.