Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 1-Pentenylboronic acid, 98%, [104376-24-1], P11073 113.951 C5H11BO2

Catalog No. 5038794
Encompass_Preferred

Compound: 1-Pentenylboronic acid

CAS#: 104376-24-1

MDL#: not listed

Background: A terminal alkenylboronic acid used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce a pentenyl group into pharmaceutical and agrochemical building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-387-94 Supplier Frontier Specialty Chemicals Supplier No. P1107325G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 1-Pentenylboronic acid

CAS#: 104376-24-1

MDL#: not listed

Background: A terminal alkenylboronic acid used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce a pentenyl group into pharmaceutical and agrochemical building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.