Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-Bromo-4-formylpyridine, 98%, [118289-17-1], B2122 186.01 C6H4BrNO

Catalog No. 50981606
Encompass_Preferred

Compound: 2-Bromo-4-formylpyridine

CAS#: 118289-17-1

MDL#: MFCD04039311

Background: 2-Bromo-4-formylpyridine is a functionalized pyridine building block whose bromine atom enables palladium-catalyzed cross-coupling reactions such as Suzuki and Stille couplings, while its aldehyde group supports nucleophilic addition, reduction, oxidation, and condensation reactions. It is a versatile intermediate for constructing substituted pyridine derivatives in pharmaceutical and agrochemical synthesis.

Catalog No. 50-981-606 Supplier Frontier Specialty Chemicals Supplier No. B212225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Bromo-4-formylpyridine

CAS#: 118289-17-1

MDL#: MFCD04039311

Background: 2-Bromo-4-formylpyridine is a functionalized pyridine building block whose bromine atom enables palladium-catalyzed cross-coupling reactions such as Suzuki and Stille couplings, while its aldehyde group supports nucleophilic addition, reduction, oxidation, and condensation reactions. It is a versatile intermediate for constructing substituted pyridine derivatives in pharmaceutical and agrochemical synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.