Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-Fluoro-5-formylphenylboronic acid, 98%, [352534-79-3], F2006 167.93 C7H6BFO3

Catalog No. 50982468
Encompass_Preferred

Compound: 2-Fluoro-5-formylphenylboronic acid

CAS#: 352534-79-3

MDL#: MFCD03095100

Background: 2-Fluoro-5-formylphenylboronic acid is a product for proteomics research and a building block chemical used in research and development. Its combination of a reactive aldehyde and boronic acid functionality makes it useful for cross-coupling and further derivatization in synthetic and pharmaceutical chemistry.

Catalog No. 50-982-468 Supplier Frontier Specialty Chemicals Supplier No. F200625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Fluoro-5-formylphenylboronic acid

CAS#: 352534-79-3

MDL#: MFCD03095100

Background: 2-Fluoro-5-formylphenylboronic acid is a product for proteomics research and a building block chemical used in research and development. Its combination of a reactive aldehyde and boronic acid functionality makes it useful for cross-coupling and further derivatization in synthetic and pharmaceutical chemistry.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.