Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-Isopropenyl boronic acid pinacol ester, 98%, [126726-62-3], I10296 168.041 C9H17BO2

Catalog No. 5037183
Encompass_Preferred

Compound: 2-Isopropenyl boronic acid pinacol ester

CAS#: 126726-62-3

MDL#: not listed

Background: An isopropenyl-substituted pinacol boronate used as a Suzuki-Miyaura coupling partner in organic synthesis. It is a versatile reagent for introducing an isopropenyl group into pharmaceutical and materials science building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-371-83 Supplier Frontier Specialty Chemicals Supplier No. I1029625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-Isopropenyl boronic acid pinacol ester

CAS#: 126726-62-3

MDL#: not listed

Background: An isopropenyl-substituted pinacol boronate used as a Suzuki-Miyaura coupling partner in organic synthesis. It is a versatile reagent for introducing an isopropenyl group into pharmaceutical and materials science building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.