Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, (2-Methylpropyl)boronic acid, 97%, [84110-40-7], I7111 101.94 C4H11BO2

Catalog No. 5037312
Encompass_Preferred

Compound: (2-Methylpropyl)boronic acid

CAS#: 84110-40-7

MDL#: not listed

Background: An isobutylboronic acid used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce a branched isobutyl group into pharmaceutical and agrochemical building blocks.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-373-12 Supplier Frontier Specialty Chemicals Supplier No. I711125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: (2-Methylpropyl)boronic acid

CAS#: 84110-40-7

MDL#: not listed

Background: An isobutylboronic acid used as a Suzuki-Miyaura coupling partner in organic synthesis. It provides a convenient means to introduce a branched isobutyl group into pharmaceutical and agrochemical building blocks.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.