Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-(Tributylstannyl)pyridine, 85%-95%, [17997-47-6], T3254 368.13 C17H31NSn

Catalog No. 50983107
Encompass_Preferred

Compound: 2-(Tributylstannyl)pyridine

CAS#: 17997-47-6

MDL#: MFCD00052052

Background: 2-(Tributylstannyl)pyridine is a nitrogen heterocycle used as a synthetic building block in pharmaceutical chemistry. It serves as an organotin reagent in Stille coupling and other transition metal-catalyzed cross-coupling reactions to form carbon-carbon bonds. This compound is classified as a dangerous good and requires hazardous shipping.

Storage: Store at 2-8°C under dry conditions.

Catalog No. 50-983-107 Supplier Frontier Specialty Chemicals Supplier No. T325425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-(Tributylstannyl)pyridine

CAS#: 17997-47-6

MDL#: MFCD00052052

Background: 2-(Tributylstannyl)pyridine is a nitrogen heterocycle used as a synthetic building block in pharmaceutical chemistry. It serves as an organotin reagent in Stille coupling and other transition metal-catalyzed cross-coupling reactions to form carbon-carbon bonds. This compound is classified as a dangerous good and requires hazardous shipping.

Storage: Store at 2-8°C under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.