Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 2-(Trifluoromethyl)pyridine-5-boronic acid, 97%, [868662-36-6], T10181 190.92 C6H5BF3NO2

Catalog No. 5039088
Encompass_Preferred

Compound: 2-(Trifluoromethyl)pyridine-5-boronic acid

CAS#: 868662-36-6

MDL#: MFCD07375381

Background: A trifluoromethylated pyridine boronic acid valued for its ability to participate in Suzuki-Miyaura coupling reactions, serving as an essential building block for pharmaceuticals, agrochemicals, and materials science research. The trifluoromethyl group enhances lipophilicity, improving the bioavailability of resulting drug candidates.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-390-88 Supplier Frontier Specialty Chemicals Supplier No. T1018125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 2-(Trifluoromethyl)pyridine-5-boronic acid

CAS#: 868662-36-6

MDL#: MFCD07375381

Background: A trifluoromethylated pyridine boronic acid valued for its ability to participate in Suzuki-Miyaura coupling reactions, serving as an essential building block for pharmaceuticals, agrochemicals, and materials science research. The trifluoromethyl group enhances lipophilicity, improving the bioavailability of resulting drug candidates.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.