Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3,3-Diethoxy-1-propenylboronic acid pinacol ester, 95%, [153737-25-8], D10252 256.146 C13H25BO4

Catalog No. 50982037
Encompass_Preferred

Compound: 3,3-Diethoxy-1-propenylboronic acid pinacol ester

CAS#: 153737-25-8

MDL#: MFCD03788737

Background: An acetal-protected propenylboronic ester used as a synthetic and pharmaceutical intermediate. It participates in Suzuki-Miyaura cross-coupling reactions, and the diethyl acetal group can be hydrolyzed after coupling to reveal an aldehyde for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-982-037 Supplier Frontier Specialty Chemicals Supplier No. D1025225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3,3-Diethoxy-1-propenylboronic acid pinacol ester

CAS#: 153737-25-8

MDL#: MFCD03788737

Background: An acetal-protected propenylboronic ester used as a synthetic and pharmaceutical intermediate. It participates in Suzuki-Miyaura cross-coupling reactions, and the diethyl acetal group can be hydrolyzed after coupling to reveal an aldehyde for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.