Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Aminophenylboronic acid monohydrate, 98%, [206658-89-1], A6087 154.96 C6H8BNO2.H2O

Catalog No. 50981070
Encompass_Preferred

Compound: 3-Aminophenylboronic acid monohydrate

CAS#: 206658-89-1

MDL#: MFCD00149554

Background: An amino-substituted phenylboronic acid used as a reagent for Suzuki-Miyaura cross-coupling. It has been used in the preparation of antivirulence drugs, boronic acid-based beta-lactamase inhibitors, and as a reversible covalent inhibitor of the dopamine transporter in pharmacological research.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-981-070 Supplier Frontier Specialty Chemicals Supplier No. A608725G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Aminophenylboronic acid monohydrate

CAS#: 206658-89-1

MDL#: MFCD00149554

Background: An amino-substituted phenylboronic acid used as a reagent for Suzuki-Miyaura cross-coupling. It has been used in the preparation of antivirulence drugs, boronic acid-based beta-lactamase inhibitors, and as a reversible covalent inhibitor of the dopamine transporter in pharmacological research.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.