Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Chloro-2-hydroxybenzaldehyde, >98%, [1927-94-2], C2016 156.57 C7H5ClO2

Catalog No. 5036440
Encompass_Preferred

Compound: 3-Chloro-2-hydroxybenzaldehyde

CAS#: 1927-94-2

MDL#: not listed

Background: A chlorinated salicylaldehyde used as a building block for Schiff base ligand synthesis in coordination chemistry, as well as pharmaceutical and agrochemical research. The phenol and aldehyde groups together enable chelation with metal ions.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-364-40 Supplier Frontier Specialty Chemicals Supplier No. C201625G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Chloro-2-hydroxybenzaldehyde

CAS#: 1927-94-2

MDL#: not listed

Background: A chlorinated salicylaldehyde used as a building block for Schiff base ligand synthesis in coordination chemistry, as well as pharmaceutical and agrochemical research. The phenol and aldehyde groups together enable chelation with metal ions.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.