Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Fluoro-2-formylpyridine, 98%, [31224-43-8], F1799 125.1 C6H4FNO

Catalog No. 5037044
Encompass_Preferred

Compound: 3-Fluoro-2-formylpyridine

CAS#: 31224-43-8

MDL#: not listed

Background: A fluorinated pyridine carbaldehyde used as a building block in pharmaceutical research. The fluorine substituent supports nucleophilic aromatic substitution chemistry, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-370-44 Supplier Frontier Specialty Chemicals Supplier No. F179925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Fluoro-2-formylpyridine

CAS#: 31224-43-8

MDL#: not listed

Background: A fluorinated pyridine carbaldehyde used as a building block in pharmaceutical research. The fluorine substituent supports nucleophilic aromatic substitution chemistry, while the aldehyde group allows for further derivatization.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.