Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, 3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester, 98%, [445303-67-3], F10178 290.06 C13H15BF4O2

Catalog No. 5036856
Encompass_Preferred

Compound: 3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester

CAS#: 445303-67-3

MDL#: MFCD09878544

Background: A fluorinated, trifluoromethylated phenylboronic ester used as a proteomics research reagent and Suzuki-Miyaura coupling partner. The multiple fluorine substituents can improve metabolic stability of downstream compounds in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-368-56 Supplier Frontier Specialty Chemicals Supplier No. F1017825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: 3-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester

CAS#: 445303-67-3

MDL#: MFCD09878544

Background: A fluorinated, trifluoromethylated phenylboronic ester used as a proteomics research reagent and Suzuki-Miyaura coupling partner. The multiple fluorine substituents can improve metabolic stability of downstream compounds in pharmaceutical research.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.